1,3,7-Triaza-spiro[4.5]decane-2,4-dione 97%

Supplier: Ambeed
Warning

A123121-250MG A123121-100MG
77189-738EA 29 USD
77189-738 77614-964
1,3,7-Triaza-spiro[4.5]decane-2,4-dione 97%
1,3,7-Triaza-spiro[4.5]decane-2,4-dione

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MDL Number: MFCD07776799
CAS Number: 78222-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR