1,4-Bis(4-aminophenoxy)benzene 99%

Supplier: Ambeed
Warning

Synonyms: 4-[4-(4-aminophenoxy)phenoxy]aniline

A122980-1G A122980-5G
77189-730EA 5.15 USD
77189-730 77745-901
1,4-Bis(4-aminophenoxy)benzene 99%
1,4-Bis(4-aminophenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O₂
MW: 292.33 g/mol
MDL Number: MFCD00039153
CAS Number: 3491-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR