tert-Butyl (5-bromopyridin-3-yl)carbamate 95%
Supplier: Ambeed
|
Warning
|
A122821-10G
A122821-1G
A122821-250MG
A122821-25G
A122821-5G
77161-418EA
88
USD
77161-418
77161-420
77161-422
77161-424
77161-426
tert-Butyl (5-bromopyridin-3-yl)carbamate 95%
tert-Butyl (5-bromopyridin-3-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrN₂O₂ MW: 273.1300 g/mol |
MDL Number:
MFCD07367900 CAS Number: 361550-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...