Methyl 2-cyano-5-fluorobenzoate 97%

Supplier: Ambeed
Warning

A122735-100G A122735-10G A122735-1G A122735-25G A122735-5G
77161-404EA 924.01 USD
77161-404 77161-406 77161-408 77161-410 77161-412
Methyl 2-cyano-5-fluorobenzoate 97%
Methyl 2-cyano-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MDL Number: MFCD11042789
CAS Number: 606080-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR