2,3-Pyridinedicarboximide 97%

Supplier: Ambeed
Warning

A122582-1G A122582-25G A122582-100G
77161-402EA 19 USD
77161-402 77566-312 77891-874
2,3-Pyridinedicarboximide 97%
2,3-Pyridinedicarboximide

Precaución: For research use only.

Formula: C₇H₄N₂O₂
MDL Number: MFCD00023018
CAS Number: 4664-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR