1-(Cyclopropylmethyl)piperazine dihydrochloride 97%

Supplier: Ambeed
Warning

A122545-100G A122545-1G A122545-250MG A122545-25G A122545-5G
77161-392EA 865.2 USD
77161-392 77161-394 77161-396 77161-398 77161-400
1-(Cyclopropylmethyl)piperazine dihydrochloride 97%
1-(Cyclopropylmethyl)piperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂
MW: 213.146 g/mol
MDL Number: MFCD08544420
CAS Number: 373608-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR