Evofosamide 98%

Supplier: Ambeed
Danger

Synonyms: TH302, TH-302, Phosphorodiamidic acid, 2-Bromo-N-[(2-bromoethylamino)-[(3-methyl-2-nitroimidazol-4-yl)methoxy]phosphoryl]ethanamine, (1-methyl-2-nitro-1H-imidazol-5-yl)methyl ester, N,N'-bis(2-bromoethyl)-, Evofosfamide

A122472-10MG A122472-1MG A122472-25MG A122472-2MG A122472-5MG A122472-100MG A122472-250MG A122472-50MG
77161-382EA 56.65 USD
77161-382 77161-384 77161-386 77161-388 77161-390 77683-098 77683-099 77683-100
Evofosamide 98%
Evofosamide

Precaución: For research use only.

Formula: C₉H₁₆Br₂N₅O₄P
MW: 449.04 g/mol
MDL Number: MFCD22420822
CAS Number: 918633-87-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR