3-Chloro-pyrazine-2-carboxylic acid ethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-chloropyrazine-2-carboxylate

A122448-1G A122448-25G A122448-5G
77161-376EA 17.51 USD
77161-376 77161-378 77161-380
3-Chloro-pyrazine-2-carboxylic acid ethyl ester 98%
3-Chloro-pyrazine-2-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.596 g/mol
MDL Number: MFCD07371661
CAS Number: 655247-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR