1,2-Dimethyl-1H-pyrrole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A122384-100MG A122384-10G A122384-1G A122384-250MG A122384-5G
77161-366EA 14 USD
77161-366 77161-368 77161-370 77161-372 77161-374
1,2-Dimethyl-1H-pyrrole-3-carboxylic acid 98%
1,2-Dimethyl-1H-pyrrole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD19230065
CAS Number: 89776-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR