4-Bromo-2,5-dimethybenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,5-dimethylphenylboronic acid, (4-Bromo-2,5-dimethylphenyl)boronic acid

A122305-1G A122305-250MG A122305-5G
77189-714EA 53.56 USD
77189-714 77189-716 77189-718
4-Bromo-2,5-dimethybenzeneboronic acid 98%
4-Bromo-2,5-dimethybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₂
MW: 228.88 g/mol
MDL Number: MFCD04115647
CAS Number: 130870-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR