6-Bromo-3-methylisoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A122287-100MG A122287-250MG A122287-1G
77161-362EA 52 USD
77161-362 77161-364 77571-352
6-Bromo-3-methylisoquinolin-1(2H)-one 95%
6-Bromo-3-methylisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD26397758
CAS Number: 872018-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR