1-Butyl-1-methylpyrrolidinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-1-methylpyrrolidin-1-ium chloride

A122253-100G A122253-10G A122253-1G
77161-356EA 216.3 USD
77161-356 77161-358 77161-360
1-Butyl-1-methylpyrrolidinium chloride 98%
1-Butyl-1-methylpyrrolidinium chloride

Precaución: for Research Use Only

Formula: C₉H₂₀ClN
MW: 177.72 g/mol
MDL Number: MFCD08458922
CAS Number: 479500-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR