4-Cyano-2-fluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A121990-100G
A121990-10G
A121990-1G
A121990-25G
A121990-5G
77189-698EA
250.01
USD
77189-698
77189-700
77189-702
77189-704
77189-706
4-Cyano-2-fluorobenzoic acid 98%
4-Cyano-2-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₄FNO₂ |
MDL Number:
MFCD03094454 CAS Number: 164149-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...