CX-5461 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-methyl-1,4-diazepan-1-yl)-N-((5-methylpyrazin-2-yl)methyl)-5-oxo-5H-benzo[4,5]thiazolo[3,2-a][1,8]naphthyridine-6-carboxamide

A121589-10MG A121589-1MG A121589-5MG A121589-250MG
77189-692EA 48.41 USD
77189-692 77189-694 77189-696 77571-288
CX-5461 ≥98%
CX-5461

Precaución: for Research Use Only

Formula: C₂₇H₂₇N₇O₂S
MW: 513.61 g/mol
MDL Number: MFCD21609261
CAS Number: 1138549-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR