1,3-Benzodioxole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A121519-100MG A121519-250MG
77189-688EA 8 USD
77189-688 77189-690
1,3-Benzodioxole-4-carboxylic acid 98%
1,3-Benzodioxole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆O₄
MDL Number: MFCD01076411
CAS Number: 5768-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR