(S)-Piperidin-3-ol 95%

Supplier: Ambeed
Warning

A121189-10G A121189-1G A121189-250MG A121189-25G A121189-5G
77161-304EA 331.66 USD
77161-304 77161-306 77161-308 77161-310 77683-088
(S)-Piperidin-3-ol 95%
(S)-Piperidin-3-ol

Precaución: For research use only.

Formula: C₅H₁₁NO
MW: 101.147 g/mol
MDL Number: MFCD08669819
CAS Number: 24211-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR