2,6-Dibromo-4-fluoropyridine 95%

Supplier: Ambeed
Warning

A121043-100MG A121043-250MG A121043-1G
77161-290EA 180.25 USD
77161-290 77161-292 77571-258
2,6-Dibromo-4-fluoropyridine 95%
2,6-Dibromo-4-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂FN
MW: 254.882 g/mol
MDL Number: MFCD13185514
CAS Number: 1214344-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR