1-Methanesulfonylpiperidin-4-ylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Methylsulphonyl)piperidin-4-amine hydrochloride

A121013-1G A121013-250MG A121013-5G
77189-678EA 11.33 USD
77189-678 77189-680 77189-682
1-Methanesulfonylpiperidin-4-ylamine hydrochloride 97%
1-Methanesulfonylpiperidin-4-ylamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₅ClN₂O₂S
MW: 214.72 g/mol
MDL Number: MFCD08690153
CAS Number: 651057-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR