CH5183284 ≥98%

Supplier: Ambeed
Warning

A120438-10MG A120438-1MG A120438-25MG A120438-50MG A120438-5MG
77189-632EA 47 USD
77189-632 77189-634 77189-636 77189-638 77189-640
CH5183284 ≥98%
CH5183284

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₆O
MDL Number: MFCD28502053
CAS Number: 1265229-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR