(R)-5-Bromo-N,N-bis-(tert-butoxycarbonyl)-3-(1-(2,6-dichloro-3-fluorophenyl)ethoxy)pyridin-2-amine 98%

Supplier: Ambeed
Warning

A120084-100MG A120084-250MG
77161-244EA 13.39 USD
77161-244 77161-246
(R)-5-Bromo-N,N-bis-(tert-butoxycarbonyl)-3-(1-(2,6-dichloro-3-fluorophenyl)ethoxy)pyridin-2-amine 98%
(R)-5-Bromo-N,N-bis-(tert-butoxycarbonyl)-3-(1-(2,6-dichloro-3-fluorophenyl)ethoxy)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₂₃H₂₆BrCl₂FN₂O₅
MW: 580.27 g/mol
MDL Number: MFCD28134573
CAS Number: 877399-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR