2-(Benzyloxy)pyridin-4-amine 97%

Supplier: Ambeed
Warning

A119963-100MG A119963-1G A119963-250MG
77161-228EA 55.62 USD
77161-228 77161-230 77161-232
2-(Benzyloxy)pyridin-4-amine 97%
2-(Benzyloxy)pyridin-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
MDL Number: MFCD16084387
CAS Number: 1214900-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR