Benzofuran-5-carbonitrile 97%

Supplier: Ambeed
Warning

A119770-100MG A119770-1G A119770-250MG A119770-5G
77189-622EA 14 USD
77189-622 77189-624 77189-626 77189-628
Benzofuran-5-carbonitrile 97%
Benzofuran-5-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅NO
MDL Number: MFCD05664402
CAS Number: 79002-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR