1-(2-Bromoethoxy)-4-fluorobenzene 98%

Supplier: Ambeed
Warning

A119760-1G A119760-250MG A119760-25G A119760-5G
77189-614EA 23 USD
77189-614 77189-616 77189-618 77189-620
1-(2-Bromoethoxy)-4-fluorobenzene 98%
1-(2-Bromoethoxy)-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MDL Number: MFCD00044739
CAS Number: 332-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR