4-N-Fmoc-aminocyclohexanone 98%

Supplier: Ambeed
Warning

Synonyms: 4-N-Fmoc-Amino-cyclohexanone

A119555-100MG A119555-1G
77161-212EA 5 USD
77161-212 77161-214
4-N-Fmoc-aminocyclohexanone 98%
4-N-Fmoc-aminocyclohexanone

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₃
MW: 335.4000 g/mol
MDL Number: MFCD03001741
CAS Number: 391248-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR