2-Ethynyl-4-fluorobenzaldehyde 97%

Supplier: Ambeed
Warning

A119318-1G A119318-250MG A119318-5G A119318-100MG
77189-564EA 243.08 USD
77189-564 77189-566 77189-568 77614-932
2-Ethynyl-4-fluorobenzaldehyde 97%
2-Ethynyl-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₅FO
MW: 148.134 g/mol
MDL Number: MFCD18824363
CAS Number: 749874-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR