(S)-tert-Butyl 3-cyanopiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A119183-100MG A119183-250MG A119183-5G
77189-554EA 22.66 USD
77189-554 77189-556 77189-558
(S)-tert-Butyl 3-cyanopiperidine-1-carboxylate 98%
(S)-tert-Butyl 3-cyanopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
MDL Number: MFCD05863903
CAS Number: 915226-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR