1,4-Di-tert-butyl-2,5-dimethoxybenzene 96%

Supplier: Ambeed
Warning

A119012-100G A119012-10G A119012-1G A119012-250MG A119012-25G A119012-5G
77189-540EA 122 USD
77189-540 77189-542 77189-544 77189-546 77189-548 77189-550
1,4-Di-tert-butyl-2,5-dimethoxybenzene 96%
1,4-Di-tert-butyl-2,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₆H₂₆O₂
MDL Number: MFCD00026281
CAS Number: 7323-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR