2,4,6-Trimethyl-2,4,6-triphenyl-1,3,5,2,4,6-trioxatrisilinane(mixture of tetramer) 97%

Supplier: Ambeed
Warning

A118935-10G A118935-5G
77189-536EA 166 USD
77189-536 77189-538
2,4,6-Trimethyl-2,4,6-triphenyl-1,3,5,2,4,6-trioxatrisilinane(mixture of tetramer) 97%
2,4,6-Trimethyl-2,4,6-triphenyl-1,3,5,2,4,6-trioxatrisilinane(mixture of tetramer)

Precaución: for Research Use Only

Formula: C₂₁H₂₄O₃Si₃
MDL Number: MFCD00053565
CAS Number: 546-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR