Ethyl-2,4-dimethylthiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,4-dimethylthiazole-5-carboxylate

A118870-10G A118870-1G A118870-25G A118870-5G
77189-522EA 62.83 USD
77189-522 77189-524 77189-526 77189-528
Ethyl-2,4-dimethylthiazole-5-carboxylate 98%
Ethyl 2,4-dimethylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂S
MW: 185.24 g/mol
MDL Number: MFCD00052874
CAS Number: 7210-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR