1-Methylpiperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A118832-100MG A118832-250MG A118832-1G
77161-198EA 42.23 USD
77161-198 77161-200 77549-382
1-Methylpiperidine-2,6-dione 97%
1-Methylpiperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MW: 127.14 g/mol
MDL Number: MFCD01725031
CAS Number: 25077-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR