Bis(4-methoxyphenyl)iodonium bromide 95%

Supplier: Ambeed
Warning

A118765-100G A118765-10G A118765-1G A118765-250MG A118765-5G
77161-176EA 611.82 USD
77161-176 77161-178 77161-180 77161-182 77161-184
Bis(4-methoxyphenyl)iodonium bromide 95%
Bis(4-methoxyphenyl)iodonium bromide

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrIO₂
MW: 421.06 g/mol
MDL Number: MFCD00031571
CAS Number: 19231-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR