5-Bromo-1-ethylpyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A118626-1G A118626-250MG
77161-172EA 32.96 USD
77161-172 77161-174
5-Bromo-1-ethylpyridin-2(1H)-one 98%
5-Bromo-1-ethylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD00456284
CAS Number: 63785-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR