2-Chloro-6,7-dimethoxy-3-methylquinoline 97%

Supplier: Ambeed
Warning

A118584-100MG A118584-1G A118584-250MG
77189-504EA 30 USD
77189-504 77189-506 77189-508
2-Chloro-6,7-dimethoxy-3-methylquinoline 97%
2-Chloro-6,7-dimethoxy-3-methylquinoline

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClNO₂
MDL Number: MFCD08899105
CAS Number: 577967-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR