Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II) 98%

Supplier: Ambeed
Warning

Synonyms: Ru(OAc)2[(R)-binap]

A118464-250MG A118464-5G A118464-1G A118464-25G
77161-164EA 15 USD
77161-164 77161-166 77610-974 77891-846
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II) 98%
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)

Precaución: for Research Use Only

Formula: C₄₈H₃₈O₄P₂Ru
MW: 841.83 g/mol
MDL Number: MFCD09753020
CAS Number: 325146-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR