3-Phenoxybenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-3-phenoxybenzene, 3-(Bromomethyl)diphenyl ether

A118387-1G A118387-250MG A118387-100MG
77189-496EA 86 USD
77189-496 77189-498 77571-096
3-Phenoxybenzyl bromide 97%
3-Phenoxybenzyl bromide

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
MW: 263.13
MDL Number: MFCD01761658
CAS Number: 51632-16-7
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR