(2-Amino-3-bromophenyl)methanol 95%

Supplier: Ambeed
Warning

A118293-1G A118293-250MG A118293-5G
77161-150EA 147.29 USD
77161-150 77161-152 77161-154
(2-Amino-3-bromophenyl)methanol 95%
(2-Amino-3-bromophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
MDL Number: MFCD18389371
CAS Number: 397323-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR