N,N'-Dicarbobenzoxy-L-cystine 95%

Supplier: Ambeed
Warning

Synonyms: (Z-Cystine-OH)₂, (Z-Cys-OH)2, N,N'-Di-Cbz-L-cystine, N,N'-Dibenzyloxycarbonyl-L-cystine

A118175-10G A118175-1G A118175-25G A118175-5G
77189-478EA 35.02 USD
77189-478 77189-480 77189-482 77189-484
N,N'-Dicarbobenzoxy-L-cystine 95%
N,N'-Dicarbobenzoxy-L-cystine

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₂O₈S₂
MW: 508.56 g/mol
MDL Number: MFCD00022030
CAS Number: 6968-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR