(S)-(-)-Tetrahydro-3-furoic acid 97%

Supplier: Ambeed
Warning

A118118-50MG A118118-1G
77189-476EA 71.07 USD
77189-476 77571-082
(S)-(-)-Tetrahydro-3-furoic acid 97%
(S)-(-)-Tetrahydro-3-furoic acid

Precaución: for Research Use Only

Formula: C₅H₈O₃
MDL Number: MFCD07369984
CAS Number: 168395-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR