2-Azaspiro[3.5]nonan-7-one, NBOC protected 95%

Supplier: Ambeed
Warning

A117777-100MG A117777-1G A117777-250MG A117777-5G A117777-25G
77161-132EA 35 USD
77161-132 77161-134 77161-136 77549-364 77891-838
2-Azaspiro[3.5]nonan-7-one, NBOC protected 95%
2-Azaspiro[3.5]nonan-7-one, NBOC protected

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₃
MDL Number: MFCD22566155
CAS Number: 1363381-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR