2-Bromo-3-chloropyridin-4-amine 95%

Supplier: Ambeed
Warning

A117742-100MG A117742-1G A117742-250MG A117742-5G
77161-124EA 117.42 USD
77161-124 77161-126 77161-128 77161-130
2-Bromo-3-chloropyridin-4-amine 95%
2-Bromo-3-chloropyridin-4-amine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MW: 207.45 g/mol
MDL Number: MFCD01646055
CAS Number: 610277-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR