N-Methylcyclobutanamine hydrochloride 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A117633-1G A117633-250MG A117633-5G A117633-100MG
77161-108EA 100 USD
77161-108 77161-110 77571-058 77683-058
N-Methylcyclobutanamine hydrochloride 95%, Ambeed.Inc
N-Methylcyclobutanamine hydrochloride

Precaución: For research use only.

Formula: C₅H₁₁N·ClH
MW: 121.608 g/mol
MDL Number: MFCD11506003
CAS Number: 848497-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR