1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 98%

Supplier: Ambeed
Warning

A117597-100G A117597-10G A117597-1G A117597-25G A117597-5G
77161-098EA 1944.64 USD
77161-098 77161-100 77161-102 77161-104 77161-106
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 98%
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₅N₃
MW: 143.15 g/mol
MDL Number: MFCD06659684
CAS Number: 517918-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR