tert-Butyl-2-ethyl-4-oxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Ethylpiperidin-4-one, N-BOC protected, 1-N-Boc-2-Ethylpiperidin-4-one

A117595-100MG A117595-1G A117595-250MG A117595-5G
77189-462EA 72.1 USD
77189-462 77189-464 77189-466 77189-468
tert-Butyl-2-ethyl-4-oxopiperidine-1-carboxylate 95%
tert-Butyl-2-ethyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
MDL Number: MFCD07367729
CAS Number: 324769-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR