tert-Butyl-3-(bromomethyl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Danger

Synonyms: 1-Boc-3-(bromomethyl)pyrrolidine

A117494-250MG A117494-1G A117494-5G
77161-096EA 18.54 USD
77161-096 77571-040 77571-042
tert-Butyl-3-(bromomethyl)pyrrolidine-1-carboxylate 97%
tert-Butyl-3-(bromomethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈BrNO₂
MW: 264.17 g/mol
MDL Number: MFCD06410515
CAS Number: 305329-97-9
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR