Ethyl 3-Oxo-3-(4-chlorophenyl)propionate 97%

Supplier: Ambeed
Warning

A117428-10G A117428-1G A117428-25G A117428-5G A117428-100G
77161-082EA 74 USD
77161-082 77161-084 77161-086 77161-088 77891-833
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate 97%
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClO₃
MDL Number: MFCD00018713
CAS Number: 2881-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR