Methyl 2-((cyanomethyl)amino)benzoate 97%

Supplier: Ambeed
Warning

A117349-100MG A117349-1G A117349-250MG
77189-450EA 69.01 USD
77189-450 77189-452 77189-454
Methyl 2-((cyanomethyl)amino)benzoate 97%
Methyl 2-((cyanomethyl)amino)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.19 g/mol
MDL Number: MFCD20661306
CAS Number: 28354-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR