Boc-HoSer-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-N-Boc-3-amino-4-hydroxybutyric acid

A117311-100MG A117311-1G A117311-250MG A117311-5G
77189-442EA 8 USD
77189-442 77189-444 77189-446 77189-448
Boc-HoSer-OH 95%
Boc-HoSer-OH

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MDL Number: MFCD00270243
CAS Number: 83345-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR