(6-Fluoroimidazo[1,2-a]pyridin-3-yl)(phenyl)methanone 97%

Supplier: Ambeed
Warning

A117256-1G A117256-250MG
77189-438EA 131.84 USD
77189-438 77189-440
(6-Fluoroimidazo[1,2-a]pyridin-3-yl)(phenyl)methanone 97%
(6-Fluoroimidazo[1,2-a]pyridin-3-yl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₉FN₂O
MW: 240.23 g/mol
MDL Number: MFCD28142906
CAS Number: 1634647-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR