1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol 97%

Supplier: Ambeed
Warning

A117246-100G A117246-10G A117246-1G A117246-5G A117246-25G
77189-430EA 38 USD
77189-430 77189-432 77189-434 77189-436 77745-867
1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol 97%
1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol

Precaución: for Research Use Only

Formula: C₂₄H₃₀O₆
MDL Number: MFCD09038711
CAS Number: 135861-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR