1-(3-Chlorophenyl)biguanide hydrochloride 97%

Supplier: Ambeed
Warning

A117131-100MG A117131-5G
77189-424EA 10 USD
77189-424 77189-426 77189-428
1-(3-Chlorophenyl)biguanide hydrochloride 97%
1-(3-Chlorophenyl)biguanide hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂N₅
MDL Number: MFCD00053019
CAS Number: 2113-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR